Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038637.1 | NP_001033726.1 | 4, 5 | 293 | ||
NM_001285908.1 | NP_001272837.1 | 3, 4 | 253 | |||
NM_001285909.1 | NP_001272838.1 | 5, 6 | 360 | |||
NM_001285910.1 | NP_001272839.1 | 2, 3 | 400 | |||
NM_001285911.1 | NP_001272840.1 | 3, 4 | 268 | |||
NM_010315.4 | NP_034445.1 | 3, 4 | 441 | |||
GenBank mRNA | AK003588.1 | - | 3, 4 | 231 | ||
AK012405.1 | - | 3, 4 | 177 | |||
AK036138.1 | - | 3, 4 | 243 | |||
AK075698.1 | - | 3, 4 | 171 | |||
AK158697.1 | - | 3, 4 | 442 | |||
AK160396.1 | - | 3, 4 | 171 | |||
BC021599.1 | - | 2, 3 | 161 | 5150203 | ||
BY108799.1 | - | 2, 3 | 163 | |||
BY245532.1 | - | 3, 4 | 442 | |||
CA324196.1 | - | 5, 6 | 283 | |||
U38496.1 | - | 1, 2 | 28 | |||
W48309.1 | - | 3, 4 | 188 |