Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080743.1 | NP_001074212.1 | 2 | 642 | ||
NM_019497.2 | NP_062370.2 | 2 | 642 | |||
GenBank mRNA | AF040745.1 | - | 2 | 68 | ||
AK020792.1 | - | 2 | 642 | |||
AK078475.1 | - | 2 | 514 | |||
AK132685.1 | - | 2 | 643 | |||
BC150691.1 | - | 2 | 166 | 9087602 | ||
BY719486.1 | - | 2 | 514 |