Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Guk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159410.1 | NP_001152882.1 | 5 | 343 | ||
NM_008193.3 | NP_032219.2 | 5 | 506 | |||
GenBank mRNA | AK004205.1 | - | 5 | 365 | ||
AK013514.1 | - | 5 | 369 | |||
BC024625.1 | - | 5 | 370 | 4485215 | ||
CO429095.1 | - | 5 | 332 | |||
EL957688.1 | - | 5 | 350 | |||
U53514.1 | - | 5 | 326 |