Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H60a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010400.2 | NP_034530.1 | 337 | |||
GenBank mRNA | AF084643.1 | - | 146 | |||
AK166223.1 | - | 294 | ||||
AK169909.1 | - | 339 | ||||
AK170137.1 | - | 310 |