Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnf4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008261.2 | NP_032287.2 | 6 | 826 | ||
GenBank mRNA | AK143948.1 | - | 6 | 851 | ||
BC039220.1 | - | 6 | 826 | 4238842 | ||
D29015.1 | - | 6 | 753 | |||
EF193391.1 | - | 6 | 625 | |||
EF193393.1 | - | 6 | 651 | |||
FJ608819.1 | - | 7 | 780 | |||
FJ608820.1 | - | 7 | 780 | |||
FJ608821.1 | - | 7 | 780 |