Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039129.4 | NP_001034218.1 | 2 | 180 | ||
NM_010447.5 | NP_034577.1 | 2 | 180 | |||
GenBank mRNA | AK007802.1 | - | 2 | 126 | ||
AK088308.1 | - | 2 | 125 | |||
AK131999.1 | - | 2 | 146 | |||
AK135391.1 | - | 2 | 124 | |||
AK136113.1 | - | 2 | 148 | |||
AK146126.1 | - | 2 | 124 | |||
AK146630.1 | - | 2 | 146 | |||
AK152684.1 | - | 2 | 124 | |||
AK161730.1 | - | 2 | 124 | |||
AK166901.1 | - | 2 | 124 | |||
AK166989.1 | - | 2 | 122 | |||
AK167161.1 | - | 2 | 124 | |||
AK167810.1 | - | 2 | 124 | |||
AK167811.1 | - | 2 | 124 | |||
AK167820.1 | - | 2 | 126 | |||
AK167913.1 | - | 2 | 124 | |||
AK168420.1 | - | 2 | 124 | |||
AK168531.1 | - | 2 | 127 | |||
AK168733.1 | - | 2 | 124 | |||
AK169216.1 | - | 2 | 124 | |||
BC052723.1 | - | 2 | 110 | |||
BC080675.1 | - | 2 | 106 | 6308746 | ||
BC083136.1 | - | 2 | 91 | 6511966 | ||
BC089340.1 | - | 2 | 99 | 6402927 | ||
BC092395.1 | - | 2 | 100 | 6403376 | ||
D86728.1 | - | 2 | 87 | |||
D86729.1 | - | 2 | 87 | |||
M99167.1 | - | 2 | 122 |