Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130419.2 | NP_001123891.1 | 3 | 601 | ||
NM_001286081.1 | NP_001273010.1 | 3 | 514 | |||
NM_001286083.1 | NP_001273012.1 | 3 | 446 | |||
NM_010471.4 | NP_034601.1 | 3 | 505 | |||
GenBank mRNA | AF326551.1 | - | 2 | 628 | ||
AF326552.1 | - | 2 | 628 | |||
AK002992.1 | - | 3 | 505 | |||
BC049607.1 | - | 3 | 470 | 6595363 | ||
BC058588.1 | - | 2 | 419 | 5360894 | ||
CX238650.1 | - | 3 | 478 | |||
CX239660.1 | - | 4 | 600 | |||
CX240088.1 | - | 3 | 446 | |||
CX240757.1 | - | 3 | 494 |