Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il2rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008368.4 | NP_032394.1 | 10 | 1711 | ||
GenBank mRNA | AK017288.1 | - | 10 | 1711 | ||
AK035289.1 | - | 10 | 1664 | |||
AK157572.1 | - | 10 | 1712 | |||
AK169481.1 | - | 10 | 1692 | |||
BC026869.1 | - | 10 | 1684 | 4015306 | ||
BC099616.1 | - | 10 | 1685 | 30024496 | ||
M28052.1 | - | 10 | 1668 |