Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008451.2 | NP_032477.2 | 13 | 1769 | ||
GenBank mRNA | AF055666.1 | - | 13 | 1599 | ||
AK141439.1 | - | 13 | 1694 | |||
BC014845.1 | - | 13 | 1613 | 2599082 | ||
CX565742.1 | - | 5 | 369 |