Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004184.3 | NP_004175.2 | 6 | 1013 | ||
NM_173701.1 | NP_776049.1 | 6 | 789 | |||
NM_213645.1 | NP_998810.1 | 5 | 617 | |||
NM_213646.1 | NP_998811.1 | 5 | 841 | |||
XM_005268044.3 | XP_005268101.1 | 6 | 741 | |||
XM_006720249.3 | XP_006720312.1 | 7 | 975 | |||
XM_011537133.2 | XP_011535435.1 | 8 | 987 | |||
XM_011537135.2 | XP_011535437.1 | 7 | 871 | |||
XM_011537136.2 | XP_011535438.1 | 6 | 707 | |||
XM_017021627.1 | XP_016877116.1 | 7 | 879 | |||
XM_017021628.1 | XP_016877117.1 | 7 | 1010 | |||
XM_017021629.1 | XP_016877118.1 | 8 | 985 | |||
GenBank mRNA | AK025487.1 | - | 4 | 417 | ||
AK056100.1 | - | 6 | 1251 | |||
AK130698.1 | - | 2 | 120 | |||
AK291141.1 | - | 5 | 593 | |||
BC017489.1 | - | 6 | 889 | 3542671 | ||
BC095453.1 | - | 7 | 900 | 30418785 | ||
BG717903.1 | - | 5 | 622 | |||
BM561448.1 | - | 637 | ||||
BX248006.1 | - | 6 | 733 | |||
M77804.1 | - | 5 | 674 | |||
X59892.1 | - | 6 | 743 | |||
X62570.1 | - | 5 | 694 |