Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008552.4 | NP_032578.2 | 9 | 1150 | ||
GenBank mRNA | AK015036.1 | - | 9 | 1141 | ||
AK030261.1 | - | 9 | 1140 | |||
AK030265.1 | - | 9 | 1151 | |||
BC120790.1 | - | 4 | 472 | 40129713 | ||
BC125537.1 | - | 5 | 535 | 40130052 | ||
BC144900.1 | - | 4 | 484 | 9053448 |