Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpdz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF326526.1 | - | 10 | 1342 | ||
AF326527.1 | - | 10 | 1342 | |||
AF326528.1 | - | 10 | 1342 | |||
AF326529.1 | - | 10 | 1342 | |||
AF326530.1 | - | 10 | 1342 | |||
AF326531.2 | - | 10 | 1342 | |||
AF326532.1 | - | 10 | 1342 | |||
AF326533.1 | - | 10 | 1342 | |||
AF326534.1 | - | 10 | 1342 | |||
AF326535.1 | - | 10 | 1342 | |||
AF326536.1 | - | 10 | 1342 | |||
AF326537.1 | - | 10 | 1342 | |||
AF326538.1 | - | 10 | 1342 | |||
AF326539.1 | - | 10 | 1342 | |||
AF326540.1 | - | 10 | 1342 | |||
AF326541.1 | - | 10 | 1342 | |||
AF326542.1 | - | 10 | 1342 | |||
AF326543.1 | - | 10 | 1342 | |||
AF326544.1 | - | 10 | 1342 | |||
AJ131869.1 | - | 11 | 1433 | |||
BC138045.1 | - | 10 | 1405 | 8861064 | ||
BC145115.1 | - | 10 | 1405 | |||
BC145117.1 | - | 10 | 1405 | 9053665 |