Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103171.1 | NP_001096641.1 | 16 | 5083 | ||
NM_010862.2 | NP_034992.2 | 16 | 5083 | |||
NM_182698.2 | NP_874357.2 | 15 | 1321 | |||
GenBank mRNA | AB014510.1 | - | 12 | 1086 | ||
AF053130.1 | - | 14 | 1289 | |||
AF144095.1 | - | 16 | 5083 | |||
AY331132.1 | - | 16 | 5083 | |||
AY331133.1 | - | 15 | 1321 |