Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008738.2 | NP_032764.1 | 3 | 718 | ||
GenBank mRNA | AK002508.1 | - | 3 | 857 | ||
BC057993.1 | - | 3 | 699 | 5345262 | ||
U78109.1 | - | 3 | 718 |