Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195046.1 | NP_001181975.1 | 14, 15 | 1545 | ||
NM_001195047.1 | NP_001181976.1 | 13, 14 | 1500 | |||
NM_001195048.1 | NP_001181977.1 | 14, 15 | 1655 | |||
NM_001195049.1 | NP_001181978.1 | 16, 17 | 1919 | |||
NM_008778.3 | NP_032804.2 | 13, 14 | 1528 | |||
GenBank mRNA | AF082297.1 | - | 12, 13 | 1442 | ||
AJ496262.1 | - | 12, 13 | 1410 | |||
AJ496263.1 | - | 13, 14 | 1455 | |||
AK031853.1 | - | 14, 15 | 1536 | |||
AK158893.1 | - | 14, 15 | 1655 | |||
AM943853.1 | - | 13, 14 | 1473 | |||
AM943854.1 | - | 14, 15 | 1518 | |||
BC053403.1 | - | 13, 14 | 1535 | 30060082 | ||
U39738.1 | - | 17, 18 | 1926 |