Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270927.1 | NP_001257856.1 | 3 | 1216 | ||
NM_001270928.1 | NP_001257857.1 | 3 | 1233 | |||
NM_001270929.1 | NP_001257858.1 | 3 | 1228 | |||
NM_001270930.1 | NP_001257859.1 | 3 | 1354 | |||
NM_001548.4 | NP_001539.3 | 2 | 1119 | |||
GenBank mRNA | AK092813.1 | - | 2 | 2147 | ||
AK308688.1 | - | 3 | 1120 | |||
AK314588.1 | - | 2 | 1042 | |||
BC007091.1 | - | 2 | 1044 | 4250452 | ||
BT006667.1 | - | 1 | 952 | |||
X03557.1 | - | 2 | 1016 |