Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270927.1 | NP_001257856.1 | 3 | 521 | ||
NM_001270928.1 | NP_001257857.1 | 3 | 538 | |||
NM_001270929.1 | NP_001257858.1 | 3 | 533 | |||
NM_001270930.1 | NP_001257859.1 | 3 | 659 | |||
NM_001548.4 | NP_001539.3 | 2 | 424 | |||
GenBank mRNA | AK092813.1 | - | 2 | 1452 | ||
AK308688.1 | - | 3 | 425 | |||
AK314588.1 | - | 2 | 347 | |||
AL548182.3 | - | 2 | 327 | |||
AL711085.1 | - | 3 | 434 | |||
BC007091.1 | - | 2 | 349 | 4250452 | ||
BI670242.1 | - | 2 | 432 | |||
BT006667.1 | - | 1 | 257 | |||
CD106320.1 | - | 2 | 300 | |||
DA822484.1 | - | 2 | 406 | |||
DB003897.1 | - | 3 | 458 | |||
DB180094.1 | - | 3 | 468 | |||
X03557.1 | - | 2 | 321 |