Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PSAP SIRNA Assays ›
Gene Symbol:
Psap
Gene Name:
prosaposin
Gene Aliases:
AI037048, SGP-1
Chromosome Location:
Chr.10: 60277628 - 60302600 on Build GRCm38
UniGene ID:
Mm.277498
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s72251
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Psap
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146120.1
NP_001139592.1
3
343
NM_001146121.1
NP_001139593.1
3
343
NM_001146122.1
NP_001139594.1
3
343
NM_001146123.1
NP_001139595.1
3
343
NM_001146124.1
NP_001139596.1
3
259
NM_011179.3
NP_035309.3
3
343
GenBank mRNA
AK088369.1
-
3
257
AK089998.1
-
3
261
AK093881.1
-
3
257
AK135637.1
-
3
257
AK145607.1
-
3
259
AK145802.1
-
3
258
AK148150.1
-
3
265
AK148152.1
-
3
284
AK148545.1
-
3
259
AK149785.1
-
3
260
AK150331.1
-
3
259
AK150645.1
-
3
257
AK151034.1
-
3
257
AK151173.1
-
3
258
AK151230.1
-
3
257
AK151234.1
-
3
259
AK151251.1
-
3
257
AK151525.1
-
3
259
AK151529.1
-
3
259
AK151686.1
-
3
293
AK151852.1
-
3
259
AK151875.1
-
3
257
AK151891.1
-
3
257
AK151907.1
-
3
259
AK151943.1
-
3
259
AK152117.1
-
3
259
AK152126.1
-
3
257
AK152134.1
-
3
259
AK152203.1
-
3
291
AK152280.1
-
3
257
AK152319.1
-
3
261
AK152355.1
-
3
259
AK152408.1
-
3
262
AK152505.1
-
3
259
AK152557.1
-
3
259
AK152605.1
-
3
259
AK152819.1
-
3
271
AK153404.1
-
3
257
AK159108.1
-
3
287
AK159164.1
-
3
257
AK159198.1
-
3
257
AK159221.1
-
3
261
AK159244.1
-
3
257
AK159266.1
-
3
257
AK159333.1
-
3
257
AK159341.1
-
3
257
AK159384.1
-
3
257
AK159414.1
-
3
260
AK159427.1
-
3
259
AK159451.1
-
3
257
AK159457.1
-
3
271
AK159463.1
-
3
259
AK159487.1
-
3
257
AK159492.1
-
3
257
AK159510.1
-
3
257
AK159528.1
-
3
257
AK159530.1
-
3
257
AK159589.1
-
3
259
AK159604.1
-
3
257
AK159622.1
-
2
149
AK159634.1
-
3
257
AK159638.1
-
3
258
AK159657.1
-
3
257
AK159664.1
-
3
257
AK159685.1
-
3
257
AK159692.1
-
3
257
AK159709.1
-
3
257
AK159720.1
-
3
257
AK159722.1
-
3
257
AK159728.1
-
3
259
AK159840.1
-
3
257
AK159861.1
-
3
257
AK159862.1
-
3
259
AK159877.1
-
3
257
AK159910.1
-
3
257
AK159915.1
-
3
257
AK159936.1
-
3
259
AK159982.1
-
3
257
AK159994.1
-
3
257
AK160015.1
-
3
257
AK160019.1
-
3
259
AK160021.1
-
3
257
AK160024.1
-
3
257
AK160046.1
-
3
257
AK160049.1
-
3
259
AK160073.1
-
3
257
AK160108.1
-
3
257
AK160877.1
-
3
260
AK161313.1
-
3
274
AK166700.1
-
3
257
AK166707.1
-
3
256
AK166744.1
-
3
255
AK166811.1
-
3
257
AK166849.1
-
3
257
AK166879.1
-
3
259
AK166909.1
-
3
257
AK166920.1
-
3
257
AK166946.1
-
3
257
AK166959.1
-
3
257
AK166984.1
-
3
257
AK167014.1
-
3
257
AK167025.1
-
3
257
AK167073.1
-
3
257
AK167099.1
-
3
260
AK167108.1
-
3
257
AK167111.1
-
3
257
AK167147.1
-
3
257
AK167198.1
-
3
259
AK167213.1
-
3
257
AK167237.1
-
3
257
AK167243.1
-
3
258
AK167252.1
-
3
259
AK167418.1
-
3
259
AK167719.1
-
3
259
AK167902.1
-
3
327
AK170416.1
-
3
256
AK180106.1
-
2
113
AK194829.1
-
2
113
AK197804.1
-
2
113
AK201095.1
-
2
113
AK203509.1
-
2
113
AK210181.1
-
2
127
BC036266.1
-
2
123
BC141089.1
-
3
256
9055701
BC145359.1
-
3
256
9053917
BY325400.1
-
3
343
S36200.1
-
3
233
S71616.1
-
3
276
U27340.1
-
3
257
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001190237, NM_001190236, NM_013013, NM_001190238, XM_006256403, and others.
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
lipid metabolic process
sphingolipid metabolic process
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
regulation of lipid metabolic process
regulation of MAPK cascade
positive regulation of MAPK cascade
developmental growth
prostate gland growth
epithelial cell differentiation involved in prostate gland development
cellular response to organic substance
negative regulation of hydrogen peroxide-induced cell death
G-protein coupled receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-f6n66:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0