Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009059.2 | NP_033085.2 | 6, 7 | 963 | ||
GenBank mRNA | AK144440.1 | - | 6, 7 | 965 | ||
BC068121.1 | - | 6, 7 | 963 | 6415928 | ||
KJ650210.1 | - | 5, 6 | 771 | |||
U54639.1 | - | 6, 7 | 892 |