Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204979.1 | NP_001191908.1 | 9 | 1360 | ||
NM_011319.3 | NP_035449.1 | 9 | 1360 | |||
GenBank mRNA | AK028358.1 | - | 9 | 1292 | ||
AK075827.1 | - | 9 | 1291 | |||
AK077309.1 | - | 9 | 1241 | |||
AK082793.1 | - | 9 | 1271 | |||
AK152150.1 | - | 9 | 1315 | |||
AK153158.1 | - | 9 | 1310 | |||
BC008612.1 | - | 9 | 1249 | 3257101 | ||
BC031932.1 | - | 9 | 1250 |