Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shbg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011367.2 | NP_035497.1 | 6 | 888 | ||
GenBank mRNA | AK010959.1 | - | 6 | 887 | ||
BC120785.1 | - | 6 | 873 | 40129708 | ||
BC120787.1 | - | 6 | 873 | 40129710 | ||
U85644.1 | - | 6 | 851 |