Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scarb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205082.1 | NP_001192011.1 | 8 | 1326 | ||
NM_001205083.1 | NP_001192012.1 | 8 | 1326 | |||
NM_016741.2 | NP_058021.1 | 8 | 1326 | |||
GenBank mRNA | AK010622.1 | - | 8 | 1251 | ||
AK028191.1 | - | 8 | 1307 | |||
AK033114.1 | - | 8 | 1251 | |||
AK145588.1 | - | 8 | 1252 | |||
AK154933.1 | - | 8 | 1318 | |||
AK158122.1 | - | 8 | 1251 | |||
AK169906.1 | - | 8 | 1302 | |||
AK171602.1 | - | 8 | 1306 | |||
AK201170.1 | - | 2 | 222 | |||
BC004656.1 | - | 8 | 1306 | 3498018 | ||
CT010222.1 | - | 8 | 1114 | |||
U37799.1 | - | 8 | 1164 |