Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113569.1 | NP_001107041.1 | 13 | 1222 | ||
NM_009295.2 | NP_033321.2 | 13 | 1217 | |||
GenBank mRNA | AF326545.1 | - | 13 | 1076 | ||
AF326563.1 | - | 13 | 1076 | |||
AK004780.1 | - | 8 | 941 | |||
AK188658.1 | - | 2 | 104 | |||
BC031728.1 | - | 13 | 1149 | 4480978 | ||
D45903.1 | - | 13 | 1111 |