Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tacc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040435.2 | NP_001035525.1 | 3 | 802 | ||
GenBank mRNA | AF093542.1 | - | 3 | 303 | ||
AF156934.1 | - | 3 | 353 | |||
AF203445.1 | - | 3 | 303 | |||
AF203446.1 | - | 3 | 303 | |||
AF247674.1 | - | 3 | 303 | |||
AK076282.1 | - | 825 | ||||
AK145556.1 | - | 3 | 363 | |||
AK160291.1 | - | 3 | 364 | |||
BC003252.1 | - | 3 | 338 | 3500413 | ||
BC055071.1 | - | 2778 | ||||
BC066184.1 | - | 3 | 338 | 30457053 | ||
BC068314.1 | - | 3 | 338 | 30461783 |