Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tacr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021382.6 | NP_067357.1 | 2, 3 | 957 | ||
GenBank mRNA | AF233341.1 | - | 2, 3 | 691 | ||
AK031898.1 | - | 2, 3 | 958 | |||
AK046424.1 | - | 2, 3 | 330 | |||
BC066845.1 | - | 2, 3 | 928 | 30436441 | ||
L27827.1 | - | 2, 3 | 385 | |||
X87823.1 | - | 2, 3 | 915 |
Set Membership: |
G-protein coupled receptor |