Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009368.3 | NP_033394.2 | 6 | 2119 | ||
GenBank mRNA | AK145304.1 | - | 6 | 2104 | ||
AK148385.1 | - | 6 | 1983 | |||
AK161328.1 | - | 3 | 366 | |||
AK162560.1 | - | 6 | 883 | |||
BC005513.1 | - | 6 | 861 | |||
BC014690.1 | - | 6 | 1398 | 3663150 | ||
BC089347.1 | - | 6 | 2076 | |||
BC094591.1 | - | 7 | 1544 | |||
BC108426.1 | - | 6 | 2119 | 6757707 | ||
M32745.1 | - | 6 | 1614 |