Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Top2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011623.2 | NP_035753.2 | 32, 33 | 4243 | ||
GenBank mRNA | AK138079.1 | - | 6, 7 | 675 | ||
D12513.1 | - | 32, 33 | 4169 | |||
U01915.1 | - | 4, 5 | 363 |