Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011675.2 | NP_035805.2 | 2 | 289 | ||
GenBank mRNA | BC025146.1 | - | 2 | 256 | 4913412 | |
BY338721.1 | - | 2 | 290 | |||
L31783.1 | - | 2 | 149 |