Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vhl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009507.3 | NP_033533.1 | 2 | 405 | ||
GenBank mRNA | AK159238.1 | - | 2 | 426 | ||
AK161408.1 | - | 2 | 402 | |||
AK171264.1 | - | 2 | 407 | |||
AK171604.1 | - | 2 | 415 | |||
BC052417.1 | - | 2 | 404 | 6402536 | ||
S76748.1 | - | 2 | 405 | |||
U12570.1 | - | 2 | 421 |