Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271757.1 | NP_001258686.1 | 4 | 578 | ||
NM_001271758.1 | NP_001258687.1 | 4 | 521 | |||
NM_009525.3 | NP_033551.2 | 4 | 775 | |||
GenBank mRNA | AK013218.1 | - | 4 | 746 | ||
AK049558.1 | - | 4 | 545 | |||
BC010775.1 | - | 4 | 521 | 4211437 | ||
BC051406.1 | - | 4 | 706 | 6438917 | ||
BU520338.1 | - | 2 | 187 | |||
M89799.1 | - | 3 | 390 |