Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF086810.1 | - | 2 | 105 | ||
AF234838.1 | - | 7 | 1493 | |||
AK140319.1 | - | 7 | 1459 | |||
AK140325.1 | - | 7 | 1459 | |||
AK155159.1 | - | 1 | 656 | |||
BC057334.1 | - | 7 | 1511 | 6811850 |