Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF074018.1 | - | 7 | 1706 | ||
AK087774.1 | - | 7 | 1728 | |||
AK135731.1 | - | 1 | 667 | |||
AY196361.1 | - | 7 | 1706 | |||
AY196362.1 | - | 7 | 1706 | |||
AY329484.1 | - | 6 | 1658 | |||
AY329485.1 | - | 7 | 1706 | |||
AY329486.1 | - | 7 | 1706 | |||
AY329487.1 | - | 6 | 1658 | |||
AY329488.1 | - | 7 | 1706 | |||
AY329489.1 | - | 7 | 1706 | |||
AY329490.1 | - | 7 | 1706 | |||
AY329491.1 | - | 7 | 1706 | |||
BC053384.1 | - | 8 | 1773 | 30049556 |