Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpdu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301710.1 | NP_001288639.1 | 2, 3 | 244 | ||
NM_001301711.1 | NP_001288640.1 | 2, 3 | 244 | |||
NM_011900.4 | NP_036030.2 | 2, 3 | 244 | |||
GenBank mRNA | AB024713.1 | - | 2, 3 | 172 | ||
AK040432.1 | - | 2, 3 | 183 | |||
AK080233.1 | - | 2, 3 | 238 | |||
AK089426.1 | - | 2, 3 | 186 | |||
AK172020.1 | - | 2, 3 | 170 | |||
BC026776.1 | - | 2, 3 | 166 | 4159355 | ||
BC048871.1 | - | 2, 3 | 172 | 5364408 | ||
CV561188.1 | - | 2, 3 | 195 | |||
CV561394.1 | - | 2, 3 | 164 |