Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Axl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190974.1 | NP_001177903.1 | 10 | 1642 | ||
NM_001190975.1 | NP_001177904.1 | 9 | 1378 | |||
NM_009465.4 | NP_033491.2 | 11 | 1669 | |||
GenBank mRNA | AK033237.1 | - | 10 | 1611 | ||
AK155567.1 | - | 12 | 1555 | |||
AK161294.1 | - | 4 | 342 | |||
AK170010.1 | - | 10 | 1616 | |||
AK170897.1 | - | 10 | 1275 | |||
BC008551.1 | - | 6 | 759 | |||
BC046618.1 | - | 11 | 1506 | 6494383 | ||
BC050914.1 | - | 11 | 1477 | 6418252 | ||
BC058230.1 | - | 11 | 1530 | 6313662 | ||
X63535.1 | - | 11 | 1418 |