Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psma6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011968.3 | NP_036098.1 | 4 | 391 | ||
GenBank mRNA | AF060087.1 | - | 4 | 300 | ||
AK088143.1 | - | 4 | 329 | |||
AK152915.1 | - | 4 | 350 | |||
AK161662.1 | - | 4 | 330 | |||
BC086685.1 | - | 4 | 318 | 30681804 | ||
U60288.1 | - | 4 | 333 |