Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081457.2 | NP_001074926.1 | 9 | 1103 | ||
NM_001081458.2 | NP_001074927.1 | 9 | 1103 | |||
NM_001135001.1 | NP_001128473.1 | 11 | 1334 | |||
NM_012023.3 | NP_036153.2 | 9 | 1103 | |||
GenBank mRNA | AB055635.1 | - | 9 | 953 | ||
AB055636.1 | - | 9 | 953 | |||
AK031584.1 | - | 9 | 1106 | |||
AK129042.1 | - | 11 | 1206 | |||
AK191098.1 | - | 4 | 319 | |||
AK196453.1 | - | 4 | 320 | |||
BC003979.1 | - | 9 | 1063 | 3489999 | ||
CN532075.1 | - | 1 | 53 | |||
CX568604.1 | - | 1 | 27 | |||
U37353.1 | - | 9 | 932 |