Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039169.1 | NP_001034258.1 | 4 | 901 | ||
NM_001039170.1 | NP_001034259.1 | 4 | 450 | |||
NM_023314.3 | NP_075803.2 | 4 | 450 | |||
GenBank mRNA | AF068116.1 | - | 4 | 364 | ||
AK017221.1 | - | 4 | 902 | |||
AK018516.1 | - | 4 | 448 | |||
AK032965.1 | - | 4 | 454 | |||
AK076067.1 | - | 2 | 165 | |||
AK138055.1 | - | 4 | 389 | |||
AK150495.1 | - | 4 | 389 | |||
BC028903.1 | - | 4 | 500 | 4166275 | ||
BC045153.1 | - | 4 | 384 | 6441327 | ||
BC049077.1 | - | 4 | 418 | 5708556 | ||
U01137.1 | - | 4 | 397 |