Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018795.2 | NP_061265.2 | 8, 9 | 994 | ||
GenBank mRNA | AB028737.1 | - | 8, 9 | 1000 | ||
BC040400.1 | - | 8, 9 | 1002 | |||
EF109740.1 | - | 8, 9 | 994 | |||
EF109741.1 | - | 8, 9 | 994 |