Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Larp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138593.2 | NP_613059.2 | 12 | 1776 | ||
GenBank mRNA | AK012726.1 | - | 1 | 78 | ||
BC029178.1 | - | 4 | 449 | |||
BC132266.1 | - | 4 | 447 | |||
BC132268.1 | - | 4 | 447 | |||
BM946683.1 | - | 6 | 644 |