Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153547.6 | NP_705775.2 | 9 | 986 | ||
GenBank mRNA | AK077523.1 | - | 9 | 917 | ||
AK167183.1 | - | 9 | 870 | |||
AY181025.1 | - | 9 | 844 | |||
AY185498.1 | - | 9 | 844 | |||
BC018560.1 | - | 9 | 825 | |||
BC037996.1 | - | 9 | 846 | 5052858 |