Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013924.3 | NP_038952.1 | 1, 2 | 290 | ||
GenBank mRNA | AB021860.1 | - | 1, 2 | 289 | ||
AK009557.1 | - | 1, 2 | 291 | |||
AK029222.1 | - | 1, 2 | 291 | |||
BC082609.1 | - | 1, 2 | 283 | 30680713 |