Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002321.1 | - | 1376 | |||
AK135614.1 | - | 1110 | ||||
AK155502.1 | - | 1380 | ||||
AK215388.1 | - | 191 | ||||
AK216099.1 | - | 194 | ||||
AK216922.1 | - | 204 | ||||
BC089604.1 | - | 1378 | 6763721 |