Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp6r3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164159.1 | NP_001157631.1 | 12 | 1550 | ||
NM_028999.1 | NP_083275.1 | 13 | 1698 | |||
NM_029456.2 | NP_083732.1 | 13 | 1698 | |||
GenBank mRNA | AK018652.1 | - | 12 | 1539 | ||
AK032179.1 | - | 4 | 469 | |||
AK035236.1 | - | 13 | 1678 | |||
AK088526.1 | - | 12 | 1518 | |||
AK129392.1 | - | 13 | 1640 | |||
AK142767.1 | - | 1 | 122 | |||
BC008529.1 | - | 13 | 1681 | 3601021 | ||
BU612294.1 | - | 1 | 92 | |||
CF727136.1 | - | 3 | 215 |