Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175095.4 | NP_780304.2 | 3 | 188 | ||
GenBank mRNA | AK047941.1 | - | 3 | 198 | ||
AK083784.1 | - | 3 | 196 | |||
AK143821.1 | - | 3 | 189 | |||
BC062098.1 | - | 175 | ||||
BC100390.1 | - | 3 | 179 | 6439921 | ||
BC119316.1 | - | 3 | 182 | 8734065 | ||
BC119318.1 | - | 3 | 182 | 8734067 |