Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002264.3 | NP_002255.3 | 11 | 1276 | ||
NR_026698.1 | 12 | 1399 | ||||
XM_005247437.3 | XP_005247494.1 | 11 | 1158 | |||
XM_005247439.3 | XP_005247496.1 | 9 | 1906 | |||
XM_011512795.2 | XP_011511097.1 | 12 | 1210 | |||
XM_011512797.2 | XP_011511099.1 | 12 | 2873 | |||
XM_017006363.1 | XP_016861852.1 | 12 | 2847 | |||
XM_017006364.1 | XP_016861853.1 | 12 | 1276 | |||
XM_017006365.1 | XP_016861854.1 | 7 | 1540 | |||
GenBank mRNA | AK125626.1 | - | 8 | 1226 | ||
AK128280.1 | - | 9 | 1906 | |||
AK299030.1 | - | 11 | 1285 | |||
AK312713.1 | - | 12 | 1394 | |||
BC002374.2 | - | 11 | 1265 | 2822859 | ||
BC003009.1 | - | 11 | 1265 | 2822859 | ||
BC090864.1 | - | 11 | 1247 | 6173605 | ||
BT006959.1 | - | 10 | 1088 | |||
CR456743.1 | - | 10 | 1088 | |||
S75295.1 | - | 11 | 1134 | |||
U20620.1 | - | 9 | 940 |