Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016804.4 | NP_058084.3 | 9 | 798 | ||
GenBank mRNA | AB041562.1 | - | 9 | 790 | ||
AF053550.1 | - | 9 | 771 | |||
AK005233.1 | - | 9 | 730 | |||
AK083055.1 | - | 760 | ||||
AK087933.1 | - | 9 | 786 | |||
AK168000.1 | - | 9 | 791 | |||
AK168021.1 | - | 9 | 761 | |||
AK168045.1 | - | 9 | 790 | |||
AK169048.1 | - | 9 | 794 | |||
AK207939.1 | - | 2 | 108 | |||
AK212612.1 | - | 2 | 108 | |||
AK216047.1 | - | 2 | 108 | |||
BC006641.1 | - | 9 | 748 | 3483318 |