Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bri3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163709.1 | NP_001157181.1 | 3 | 511 | ||
NM_018772.4 | NP_061242.3 | 3 | 627 | |||
GenBank mRNA | AF106967.1 | - | 3 | 546 | ||
AK080467.1 | - | 3 | 617 | |||
BC005677.1 | - | 3 | 605 | |||
BC021149.1 | - | 3 | 573 | |||
BC095955.1 | - | 3 | 582 | 5328625 | ||
X61454.1 | - | 2 | 275 |