Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204165.1 | NP_001191094.1 | 31 | 3043 | ||
NM_001204166.1 | NP_001191095.1 | 28 | 2935 | |||
NM_019688.2 | NP_062662.1 | 30 | 2989 | |||
GenBank mRNA | AB021132.1 | - | 31 | 3018 | ||
AB037668.1 | - | 22 | 2378 | |||
AB445011.1 | - | 28 | 2927 | |||
AF115480.1 | - | 30 | 2917 | |||
AK004874.1 | - | 22 | 2035 | |||
AK220281.1 | - | 31 | 3043 | |||
BC139822.1 | - | 31 | 2975 | 40141811 |