Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB038560.1 | - | 7 | 3501 | ||
AK013913.1 | - | 1 | 52 | |||
AK050819.1 | - | 8 | 2842 | |||
AK220315.1 | - | 7 | 3449 | |||
BC025857.1 | - | 2 | 982 | |||
BC036977.1 | - | 8 | 2774 | |||
BC092394.1 | - | 7 | 3519 | 30534262 | ||
BC141058.1 | - | 7 | 3457 | 9055670 |