Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021480.5 | NP_067455.5 | 7 | 1067 | ||
GenBank mRNA | AF134346.2 | - | 7 | 731 | ||
AK077571.1 | - | 7 | 742 | |||
AY116662.1 | - | 6 | 668 | |||
BC058860.1 | - | 7 | 1082 | 30014660 |